Assay Details
| Gene Symbol: MUC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279045.2 | XP_015134531.1 | 6 - 7 | 2261 | 64 | |
| XM_015279046.2 | XP_015134532.2 | 7 - 8 | 1270 | 64 | ||
| GenBank mRNA | CR391212.1 | - | 1 - 2 | 78 | 64 | |
More Information
Set Membership: |
|

MUC1
Genomic Map