Assay Details
Gene Symbol: C14H16orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036657.2 | 5 - 6 | 878 | 103 | ||
GenBank mRNA | BX930203.1 | - | 5 - 6 | 477 | 103 | |
BX930271.1 | - | 5 - 6 | 878 | 103 | ||
BX931816.1 | - | 1 - 2 | 451 | 103 | ||
BX932634.1 | - | 3 - 4 | 241 | 103 | ||
BX933532.2 | - | 4 - 5 | 442 | 103 | ||
BX934307.2 | - | 5 - 6 | 393 | 103 | ||
CR391334.1 | - | 2 - 3 | 224 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
