Assay Details
| Gene Symbol: CTDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418905.6 | XP_418905.5 | 10 - 11 | 2633 | 82 | |
| GenBank mRNA | CR391437.1 | - | 1 - 2 | 96 | 82 | |
More Information
Set Membership: |
|

CTDP1
Genomic Map