Assay Details
| Gene Symbol: SEC61A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321620.1 | NP_001308549.1 | 4 - 5 | 265 | 73 | |
| XM_015297820.2 | XP_015153306.1 | 5 - 6 | 398 | 73 | ||
| GenBank mRNA | CR405768.1 | - | 4 - 5 | 272 | 73 | |
| DR427846.1 | - | 4 - 5 | 265 | 73 | ||
More Information
Set Membership: |
|

SEC61A2
Genomic Map