Assay Details
Gene Symbol: TIPRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416593.6 | XP_416593.1 | 1 - 2 | 254 | 69 | |
GenBank mRNA | BX935082.2 | - | 1 - 2 | 224 | 69 | |
BX950267.2 | - | 1 - 2 | 202 | 69 | ||
CR406044.1 | - | 1 - 2 | 220 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
