Assay Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419986.5 | XP_419986.1 | 3 - 4 | 403 | 82 | |
| GenBank mRNA | BX931493.1 | - | 3 - 4 | 386 | 82 | |
| BX933820.1 | - | 3 - 4 | 376 | 82 | ||
| BX936239.1 | - | 3 - 4 | 380 | 82 | ||
| BX950504.1 | - | 3 - 4 | 418 | 82 | ||
| CR406049.1 | - | 3 - 4 | 384 | 82 | ||
More Information
Set Membership: |
|

SF3B6
Genomic Map