Assay Details
| Gene Symbol: MPHOSPH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277139.1 | NP_001264068.1 | 2 - 3 | 251 | 120 | |
| GenBank mRNA | BX929849.2 | - | 2 - 3 | 144 | 120 | |
| BX930843.2 | - | 2 - 3 | 160 | 120 | ||
| BX935551.1 | - | 2 - 3 | 250 | 120 | ||
| BX936086.2 | - | 2 - 3 | 219 | 120 | ||
| CR406488.1 | - | 2 - 3 | 144 | 120 | ||
More Information
Set Membership: |
|

MPHOSPH6
Genomic Map