Assay Details
| Gene Symbol: PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369063.1 | NP_001355992.1 | 5 - 6 | 1712 | 140 | |
| GenBank mRNA | CR390934.1 | - | 5 - 6 | 1314 | 140 | |
| CR407066.1 | - | 1 - 2 | 690 | 140 | ||
More Information
Set Membership: |
|

PTCD1
Genomic Map