Assay Details
| Gene Symbol: TAF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244905.1 | NP_001231834.1 | 351 | 74 | ||
| GenBank mRNA | CR354169.1 | - | 92 | 74 | ||
| CR523581.1 | - | 151 | 74 | |||
More Information
Set Membership: |
|

TAF11
Genomic Map