Assay Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167761.1 | NP_001161233.1 | 2 - 3 | 278 | 75 | |
| GenBank mRNA | D26093.1 | - | 2 - 3 | 279 | 75 | |
More Information
Set Membership: |
|

VMO1
Genomic Map