Assay Details
| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276479.2 | XP_015131965.1 | 4 - 5 | 710 | 62 | |
| XM_015276480.1 | XP_015131966.1 | 5 - 6 | 892 | 62 | ||
| GenBank mRNA | EU275768.1 | - | 4 - 5 | 711 | 62 | |
| MG066596.1 | - | 5 - 6 | 722 | 62 | ||
More Information
Set Membership: |
|

SCD5
Genomic Map