Assay Details
| Gene Symbol: PTGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167718.1 | NP_001161190.1 | 3 - 4 | 811 | 93 | |
| NM_001167719.1 | NP_001161191.1 | 3 - 4 | 382 | 93 | ||
| GenBank mRNA | CF250042.1 | - | 3 - 4 | 382 | 93 | |
| M64990.1 | - | 3 - 4 | 907 | 93 | ||
More Information
Set Membership: |
|

PTGS2
Genomic Map