Assay Details
| Gene Symbol: ALG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199825.1 | NP_001186754.1 | 2 - 3 | 287 | 83 | |
| GenBank mRNA | BX929850.1 | - | 2 - 3 | 287 | 83 | |
| BX930990.2 | - | 2 - 3 | 285 | 83 | ||
More Information
Set Membership: |
|

ALG11
Genomic Map