Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277918.1 | NP_001264847.1 | 4 - 5 | 481 | 79 | |
| GenBank mRNA | BU264806.1 | - | 4 - 5 | 410 | 79 | |
| BX930160.1 | - | 4 - 5 | 389 | 79 | ||
| BX933062.1 | - | 4 - 5 | 351 | 79 | ||
More Information
Set Membership: |
|

HDDC2
Genomic Map