Assay Details
Gene Symbol: C14H16orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036657.2 | 6 - 7 | 1087 | 56 | ||
GenBank mRNA | BX930203.1 | - | 6 - 7 | 685 | 56 | |
BX930271.1 | - | 6 - 7 | 1087 | 56 | ||
BX931816.1 | - | 2 - 3 | 660 | 56 | ||
BX932634.1 | - | 4 - 5 | 450 | 56 | ||
BX932942.1 | - | 5 - 6 | 723 | 56 | ||
BX933532.2 | - | 5 - 6 | 651 | 56 | ||
BX934307.2 | - | 6 - 7 | 602 | 56 | ||
BX934976.2 | - | 7 - 8 | 1055 | 56 | ||
CR352554.1 | - | 2 - 3 | 683 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
