Assay Details
| Gene Symbol: NINJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277877.1 | NP_001264806.1 | 2 - 3 | 355 | 91 | |
| GenBank mRNA | BX930736.1 | - | 2 - 3 | 456 | 91 | |
| BX935591.1 | - | 2 - 3 | 355 | 91 | ||
More Information
Set Membership: |
|

NINJ1
Genomic Map