Assay Details
| Gene Symbol: NIFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422088.5 | XP_422088.1 | 5 - 6 | 676 | 76 | |
| GenBank mRNA | BX930757.1 | - | 5 - 6 | 655 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NIFK| Gene Symbol: NIFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422088.5 | XP_422088.1 | 5 - 6 | 676 | 76 | |
| GenBank mRNA | BX930757.1 | - | 5 - 6 | 655 | 76 | |
Set Membership: |
|