Assay Details
| Gene Symbol: TSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277169.1 | NP_001264098.1 | 2 - 3 | 370 | 76 | |
| GenBank mRNA | BX930961.1 | - | 2 - 3 | 350 | 76 | |
| BX931234.1 | - | 2 - 3 | 339 | 76 | ||
| BX932485.1 | - | 2 - 3 | 330 | 76 | ||
| CR733399.1 | - | 2 - 3 | 306 | 76 | ||
More Information
Set Membership: |
|

TSR3
Genomic Map