Assay Details
| Gene Symbol: GLT8D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198607.1 | NP_001185536.1 | 4 - 5 | 601 | 81 | |
| GenBank mRNA | BX931175.1 | - | 4 - 5 | 595 | 81 | |
| BX933990.2 | - | 4 - 5 | 601 | 81 | ||
| BX934022.2 | - | 4 - 5 | 578 | 81 | ||
More Information
Set Membership: |
|

GLT8D1
Genomic Map