Assay Details
| Gene Symbol: MTHFS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277626.1 | NP_001264555.1 | 1 - 2 | 161 | 80 | |
| XM_025153842.1 | XP_025009610.1 | 1 - 2 | 159 | 80 | ||
| GenBank mRNA | BX931484.1 | - | 1 - 2 | 161 | 80 | |
| BX932260.1 | - | 1 - 2 | 127 | 80 | ||
| BX934466.2 | - | 1 - 2 | 122 | 80 | ||
More Information
Set Membership: |
|

MTHFS
Genomic Map