Assay Details
| Gene Symbol: MTHFS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277626.1 | NP_001264555.1 | 2 - 3 | 422 | 85 | |
| XM_025153842.1 | XP_025009610.1 | 2 - 3 | 420 | 85 | ||
| XM_025153843.1 | XP_025009611.1 | 2 - 3 | 379 | 85 | ||
| GenBank mRNA | BX931484.1 | - | 2 - 3 | 422 | 85 | |
| BX932260.1 | - | 2 - 3 | 388 | 85 | ||
| BX934466.2 | - | 2 - 3 | 383 | 85 | ||
More Information
Set Membership: |
|

MTHFS
Genomic Map