Assay Details
| Gene Symbol: PUSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277883.1 | NP_001264812.1 | 5 - 6 | 656 | 81 | |
| NM_001277884.1 | NP_001264813.1 | 5 - 6 | 707 | 81 | ||
| XM_015296879.2 | XP_015152365.1 | 5 - 6 | 669 | 81 | ||
| XM_025142514.1 | XP_024998282.1 | 5 - 6 | 641 | 81 | ||
| XM_025142515.1 | XP_024998283.1 | 5 - 6 | 655 | 81 | ||
| GenBank mRNA | BX929622.1 | - | 5 - 6 | 707 | 81 | |
| BX931895.1 | - | 5 - 6 | 656 | 81 | ||
More Information
Set Membership: |
|

PUSL1
Genomic Map