Assay Details
| Gene Symbol: HMBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947916.3 | XP_004947973.1 | 5 - 6 | 393 | 65 | |
| XM_015298075.2 | XP_015153561.1 | 8 - 9 | 486 | 65 | ||
| XM_025143289.1 | XP_024999057.1 | 7 - 8 | 557 | 65 | ||
| XM_025143290.1 | XP_024999058.1 | 8 - 9 | 504 | 65 | ||
| XM_417846.6 | XP_417846.2 | 7 - 8 | 443 | 65 | ||
| GenBank mRNA | BX932073.1 | - | 7 - 8 | 425 | 65 | |
More Information
Set Membership: |
|

HMBS
Genomic Map