Assay Details
| Gene Symbol: ECHDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198652.3 | NP_001185581.1 | 3 - 4 | 399 | 95 | |
| GenBank mRNA | BX931998.1 | - | 3 - 4 | 354 | 95 | |
| BX932081.1 | - | 3 - 4 | 400 | 95 | ||
| BX935838.2 | - | 3 - 4 | 343 | 95 | ||
| CR353437.1 | - | 2 - 3 | 360 | 95 | ||
More Information
Set Membership: |
|

ECHDC3
Genomic Map