Assay Details
| Gene Symbol: ECHDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198652.3 | NP_001185581.1 | 4 - 5 | 603 | 57 | |
| GenBank mRNA | BX931998.1 | - | 4 - 5 | 558 | 57 | |
| BX932081.1 | - | 4 - 5 | 604 | 57 | ||
| BX935838.2 | - | 4 - 5 | 547 | 57 | ||
| CR353437.1 | - | 3 - 4 | 564 | 57 | ||
More Information
Set Membership: |
|

ECHDC3
Genomic Map