Assay Details
| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424908.6 | XP_424908.2 | 7 - 8 | 914 | 114 | |
| GenBank mRNA | BX932244.1 | - | 6 - 7 | 607 | 114 | |
| BX935845.2 | - | 7 - 8 | 872 | 114 | ||
More Information
Set Membership: |
|

CCNH
Genomic Map