Assay Details
| Gene Symbol: PLA2G10L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938210.3 | XP_004938267.1 | 4 - 5 | 767 | 66 | |
| XM_025143691.1 | XP_024999459.1 | 3 - 4 | 495 | 66 | ||
| GenBank mRNA | BX932556.1 | - | 2 - 3 | 401 | 66 | |
| BX934166.2 | - | 3 - 4 | 467 | 66 | ||
| CR405760.1 | - | 2 - 3 | 444 | 66 | ||
More Information
Set Membership: |
|

PLA2G10L
Genomic Map