Assay Details
| Gene Symbol: RBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277417.1 | NP_001264346.1 | 1 - 2 | 127 | 101 | |
| GenBank mRNA | BU296446.1 | - | 1 - 2 | 98 | 101 | |
| BX930467.1 | - | 1 - 2 | 120 | 101 | ||
| BX930583.2 | - | 1 - 2 | 118 | 101 | ||
| BX932676.1 | - | 1 - 2 | 137 | 101 | ||
More Information
Set Membership: |
|

RBP2
Genomic Map