Assay Details
| Gene Symbol: EMC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421207.6 | XP_421207.3 | 2 - 3 | 371 | 88 | |
| GenBank mRNA | BX932678.1 | - | 2 - 3 | 372 | 88 | |
| CR353992.2 | - | 2 - 3 | 428 | 88 | ||
| CR354308.1 | - | 2 - 3 | 393 | 88 | ||
| CR405723.1 | - | 2 - 3 | 430 | 88 | ||
More Information
Set Membership: |
|

EMC7
Genomic Map