Assay Details
| Gene Symbol: ATP6V1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422192.4 | XP_422192.1 | 1 - 2 | 238 | 62 | |
| GenBank mRNA | BX932779.1 | - | 1 - 2 | 245 | 62 | |
More Information
Set Membership: |
|

ATP6V1G3
Genomic Map