Assay Details
| Gene Symbol: CREBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278110.3 | NP_001265039.1 | 2 - 3 | 331 | 82 | |
| GenBank mRNA | BU339707.1 | - | 2 - 3 | 332 | 82 | |
| BX932584.1 | - | 2 - 3 | 330 | 82 | ||
| BX932597.1 | - | 2 - 3 | 299 | 82 | ||
| BX933150.1 | - | 2 - 3 | 298 | 82 | ||
| BX933493.2 | - | 2 - 3 | 320 | 82 | ||
| CR386358.1 | - | 2 - 3 | 317 | 82 | ||
More Information
Set Membership: |
|

CREBL2
Genomic Map