Assay Details
| Gene Symbol: RABL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278112.1 | NP_001265041.1 | 1 - 2 | 150 | 75 | |
| GenBank mRNA | BU315783.1 | - | 1 - 2 | 120 | 75 | |
| BX930967.1 | - | 1 - 2 | 130 | 75 | ||
| BX933167.1 | - | 1 - 2 | 120 | 75 | ||
| CN224784.1 | - | 1 - 2 | 150 | 75 | ||
More Information
Set Membership: |
|

RABL2A
Genomic Map