Assay Details
| Gene Symbol: RABL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278112.1 | NP_001265041.1 | 5 - 6 | 454 | 69 | |
| XM_025151044.1 | XP_025006812.1 | 3 - 4 | 423 | 69 | ||
| GenBank mRNA | BU315783.1 | - | 5 - 6 | 424 | 69 | |
| BX930967.1 | - | 5 - 6 | 434 | 69 | ||
| BX933167.1 | - | 5 - 6 | 424 | 69 | ||
| BX950545.1 | - | 1 - 2 | 497 | 69 | ||
More Information
Set Membership: |
|

RABL2A
Genomic Map