Assay Details
| Gene Symbol: RABL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278112.1 | NP_001265041.1 | 7 - 8 | 632 | 108 | |
| XM_025151044.1 | XP_025006812.1 | 5 - 6 | 601 | 108 | ||
| GenBank mRNA | BU315783.1 | - | 7 - 8 | 602 | 108 | |
| BX930967.1 | - | 7 - 8 | 612 | 108 | ||
| BX933167.1 | - | 7 - 8 | 602 | 108 | ||
| BX950545.1 | - | 3 - 4 | 675 | 108 | ||
More Information
Set Membership: |
|

RABL2A
Genomic Map