Assay Details
| Gene Symbol: SIRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276364.1 | NP_001263293.1 | 6 - 7 | 721 | 61 | |
| NR_075081.1 | 4 - 5 | 407 | 61 | |||
| XM_015275872.2 | XP_015131358.1 | 6 - 7 | 810 | 61 | ||
| XM_015275873.2 | XP_015131359.1 | 6 - 7 | 819 | 61 | ||
| XM_025147257.1 | XP_025003025.1 | 6 - 7 | 3037 | 61 | ||
| GenBank mRNA | BX933404.1 | - | 6 - 7 | 730 | 61 | |
| KX400578.1 | - | 5 - 6 | 615 | 61 | ||
More Information
Set Membership: |
|

SIRT5
Genomic Map