Assay Details
| Gene Symbol: MRPL47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422785.6 | XP_422785.2 | 1 - 2 | 160 | 95 | |
| GenBank mRNA | BX933605.1 | - | 1 - 2 | 99 | 95 | |
| BX933613.2 | - | 1 - 2 | 116 | 95 | ||
| BX934647.1 | - | 1 - 2 | 126 | 95 | ||
| BX936225.2 | - | 1 - 2 | 116 | 95 | ||
More Information
Set Membership: |
|

MRPL47
Genomic Map