Assay Details
| Gene Symbol: DCAF4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941670.3 | XP_004941727.1 | 5 - 6 | 565 | 63 | |
| GenBank mRNA | BX933708.1 | - | 5 - 6 | 583 | 63 | |
| CR405806.1 | - | 5 - 6 | 562 | 63 | ||
More Information
Set Membership: |
|

DCAF4L2
Genomic Map