Assay Details
| Gene Symbol: GCHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199522.1 | NP_001186451.1 | 2 - 3 | 149 | 73 | |
| GenBank mRNA | BX933750.1 | - | 2 - 3 | 149 | 73 | |
| BX950302.1 | - | 2 - 3 | 127 | 73 | ||
More Information
Set Membership: |
|

GCHFR
Genomic Map