Assay Details
| Gene Symbol: CZH5ORF42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX933956.1 | - | 2 - 3 | 831 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CZH5ORF42| Gene Symbol: CZH5ORF42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX933956.1 | - | 2 - 3 | 831 | 132 | |
Set Membership: |
|