Assay Details
| Gene Symbol: CTSV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168009.1 | NP_001161481.1 | 5 - 6 | 698 | 125 | |
| GenBank mRNA | BX932251.2 | - | 5 - 6 | 684 | 125 | |
| BX933970.1 | - | 5 - 6 | 685 | 125 | ||
| BX936191.1 | - | 5 - 6 | 672 | 125 | ||
More Information
Set Membership: |
|

CTSV
Genomic Map