Assay Details
| Gene Symbol: C17ORF80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415700.6 | XP_415700.4 | 3 - 4 | 1348 | 71 | |
| GenBank mRNA | BX929428.2 | - | 1 - 2 | 108 | 71 | |
| BX931333.2 | - | 3 - 4 | 400 | 71 | ||
| BX932338.2 | - | 2 - 3 | 300 | 71 | ||
| BX934155.1 | - | 1 - 2 | 35 | 71 | ||
More Information
Set Membership: |
|

C17ORF80
Genomic Map