Assay Details
| Gene Symbol: PIGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290426.2 | XP_015145912.1 | 2 | 1374 | 94 | |
| XM_422231.6 | XP_422231.3 | 2 | 1118 | 94 | ||
| GenBank mRNA | BX934391.1 | - | 1 | 1051 | 94 | |
More Information
Set Membership: |
|

PIGC
Genomic Map