Assay Details
| Gene Symbol: RAB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936788.3 | XP_004936845.1 | 4 - 5 | 490 | 99 | |
| XM_015291222.2 | XP_015146708.1 | 3 - 4 | 625 | 99 | ||
| XM_015291223.2 | XP_015146709.1 | 3 - 4 | 538 | 99 | ||
| XM_422470.6 | XP_422470.2 | 3 - 4 | 478 | 99 | ||
| GenBank mRNA | BX931637.2 | - | 2 - 3 | 145 | 99 | |
| BX934582.1 | - | 3 - 4 | 523 | 99 | ||
More Information
Set Membership: |
|

RAB3B
Genomic Map