Assay Details
| Gene Symbol: IGSF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318422.1 | NP_001305351.1 | 2 - 3 | 318 | 67 | |
| XM_015294662.2 | XP_015150148.1 | 2 - 3 | 333 | 67 | ||
| GenBank mRNA | AY332741.1 | - | 2 - 3 | 318 | 67 | |
| BX934710.1 | - | 2 - 3 | 206 | 67 | ||
More Information
Set Membership: |
|

IGSF11
Genomic Map