Assay Details
| Gene Symbol: RIDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234688.5 | XP_001234689.4 | 5 - 6 | 384 | 68 | |
| GenBank mRNA | BX932754.1 | - | 5 - 6 | 376 | 68 | |
| BX935048.1 | - | 5 - 6 | 358 | 68 | ||
More Information
Set Membership: |
|

RIDA
Genomic Map