Assay Details
Gene Symbol: EGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421233.5 | XP_421233.3 | 1 - 2 | 682 | 73 | |
GenBank mRNA | BX934904.1 | - | 1 - 2 | 657 | 73 | |
BX935456.1 | - | 1 - 2 | 687 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
