Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149345.1 | XP_025005113.1 | 4 - 5 | 535 | 62 | |
| XM_419728.6 | XP_419728.3 | 5 - 6 | 666 | 62 | ||
| GenBank mRNA | BX935610.1 | - | 4 - 5 | 699 | 62 | |
More Information
Set Membership: |
|

MTFR2
Genomic Map