Assay Details
| Gene Symbol: SEC11C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424458.6 | XP_424458.4 | 2 - 3 | 220 | 56 | |
| GenBank mRNA | BX932058.1 | - | 2 - 3 | 224 | 56 | |
| BX935228.1 | - | 2 - 3 | 203 | 56 | ||
| BX935949.2 | - | 1 - 2 | 397 | 56 | ||
| BX950629.1 | - | 2 - 3 | 237 | 56 | ||
| CR353329.2 | - | 2 - 3 | 230 | 56 | ||
More Information
Set Membership: |
|

SEC11C
Genomic Map