Assay Details
Gene Symbol: VDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015302414.2 | XP_015157900.1 | 5 - 6 | 422 | 65 | |
XM_015302417.2 | XP_015157903.1 | 6 - 7 | 467 | 65 | ||
XM_015302418.2 | XP_015157904.1 | 5 - 6 | 519 | 65 | ||
XM_015302420.2 | XP_015157906.1 | 5 - 6 | 344 | 65 | ||
XM_015302421.2 | XP_015157907.1 | 5 - 6 | 369 | 65 | ||
XM_015302422.2 | XP_015157908.1 | 6 - 7 | 402 | 65 | ||
XM_015302423.2 | XP_015157909.1 | 4 - 5 | 346 | 65 | ||
XM_015302424.2 | XP_015157910.1 | 4 - 5 | 366 | 65 | ||
XM_015302425.1 | XP_015157911.1 | 4 - 5 | 316 | 65 | ||
XM_025142891.1 | XP_024998659.1 | 6 - 7 | 469 | 65 | ||
XM_025142892.1 | XP_024998660.1 | 5 - 6 | 480 | 65 | ||
XM_025142893.1 | XP_024998661.1 | 5 - 6 | 477 | 65 | ||
XM_025142895.1 | XP_024998663.1 | 6 - 7 | 451 | 65 | ||
XM_025142896.1 | XP_024998664.1 | 5 - 6 | 476 | 65 | ||
GenBank mRNA | BX950721.1 | - | 5 - 6 | 554 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
