Assay Details
| Gene Symbol: TRMT10C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297190.2 | XP_015152676.1 | 1 - 2 | 132 | 100 | |
| GenBank mRNA | CR352607.1 | - | 1 - 2 | 113 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015297190.2 transcripts.
Set Membership: |
|

TRMT10C
Genomic Map